siRNA Details
| Gene Symbol: Ncstn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021607.3 | NP_067620.3 | 15, 16 | 1865 | ||
| GenBank mRNA | AF240469.1 | - | 15, 16 | 1815 | ||
| AK033314.1 | - | 15, 16 | 1808 | |||
| AK129102.1 | - | 15, 16 | 1865 | |||
| AK160338.1 | - | 411 | ||||
| AK170369.1 | - | 15, 16 | 1854 | |||
| AK172579.1 | - | 15, 16 | 1810 | |||
| BC019998.1 | - | 15, 16 | 1810 | 4458780 | ||

Ncstn
Genomic Map