siRNA Details
| Gene Symbol: Gpa33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021610.1 | NP_067623.1 | 5 | 719 | ||
| GenBank mRNA | AF247659.1 | - | 5 | 719 | ||
| BC008528.1 | - | 5 | 712 | 3593917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpa33| Gene Symbol: Gpa33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021610.1 | NP_067623.1 | 5 | 719 | ||
| GenBank mRNA | AF247659.1 | - | 5 | 719 | ||
| BC008528.1 | - | 5 | 712 | 3593917 | ||