siRNA Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021713.2 | NP_068359.1 | 7 | 1159 | ||
| GenBank mRNA | AF289484.1 | - | 7 | 1151 | ||
| AK002656.1 | - | 7 | 1133 | |||
| AK004104.1 | - | 7 | 1134 | |||
| AK013232.1 | - | 7 | 1105 | |||
| BC023124.1 | - | 6 | 1286 | |||
| BC028501.1 | - | 7 | 1144 | 5256915 | ||
| BU851558.1 | - | 1 | 94 | |||

Myg1
Genomic Map