siRNA Details
| Gene Symbol: Acss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019811.3 | NP_062785.2 | 9 | 1190 | ||
| GenBank mRNA | AF216873.1 | - | 9 | 1140 | ||
| AK028586.1 | - | 9 | 1168 | |||
| AK029014.1 | - | 9 | 1158 | |||
| AK029262.1 | - | 1176 | ||||
| AK053877.1 | - | 9 | 1164 | |||
| AK093831.1 | - | 9 | 1167 | |||
| BC040790.1 | - | 9 | 1140 | |||
| BC051432.1 | - | 9 | 1190 | 6515568 | ||

Acss2
Genomic Map