siRNA Details
| Gene Symbol: Acss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019811.3 | NP_062785.2 | 11 | 1452 | ||
| GenBank mRNA | AF216873.1 | - | 11 | 1402 | ||
| AK028586.1 | - | 11 | 1430 | |||
| AK029014.1 | - | 11 | 1420 | |||
| AK029262.1 | - | 1438 | ||||
| AK053877.1 | - | 11 | 1426 | |||
| AK093831.1 | - | 11 | 1429 | |||
| BC040790.1 | - | 11 | 1402 | |||
| BC051432.1 | - | 11 | 1452 | 6515568 | ||

Acss2
Genomic Map