siRNA Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021896.5 | NP_068696.2 | 6 | 1540 | ||
| GenBank mRNA | AF297082.1 | - | 6 | 1515 | ||
| AK004815.1 | - | 6 | 1474 | |||
| AK028558.1 | - | 6 | 1463 | |||
| BC057327.1 | - | 6 | 1450 | 6825960 | ||
| BC094310.1 | - | 6 | 1596 | 6838836 | ||

Gucy1a3
Genomic Map