siRNA Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038699.2 | NP_001033788.1 | 1 | 155 | ||
| NM_022014.4 | NP_071297.1 | 1 | 155 | |||
| GenBank mRNA | AA058133.1 | - | 1 | 111 | ||
| AJ404616.1 | - | 1 | 141 | |||
| AK010151.1 | - | 1 | 149 | |||
| AK172235.1 | - | 1 | 140 | |||
| BC054374.1 | - | 1 | 136 | 6313660 | ||
| BC064710.1 | - | 1 | 141 | 30380196 | ||
| BY142714.1 | - | 1 | 157 | |||

Fn3k
Genomic Map