siRNA Details
| Gene Symbol: Impg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022016.3 | NP_071299.3 | 12, 13 | 1430 | ||
| GenBank mRNA | AF229929.1 | - | 12, 13 | 1341 | ||
| AF266478.1 | - | 12, 13 | 1476 | |||
| BC022970.1 | - | 12, 13 | 1418 | 5366008 | ||

Impg1
Genomic Map