siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022018.3 | NP_071301.2 | 11 | 1582 | ||
| GenBank mRNA | AB049355.1 | - | 11 | 1422 | ||
| AK029847.1 | - | 7 | 894 | |||
| AK136597.1 | - | 11 | 1492 | |||
| AK170193.1 | - | 11 | 1505 | |||
| AK170347.1 | - | 2 | 177 | |||
| AK210084.1 | - | 1 | 71 | |||
| BC021332.1 | - | 11 | 1470 | |||
| BC038006.1 | - | 11 | 1438 | |||
| BC058234.1 | - | 9 | 1182 | 6306849 | ||
| BC099690.1 | - | 11 | 1489 | |||
| BC137843.1 | - | 7 | 959 | 8860862 | ||

Fam129a
Genomic Map