siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022018.3 | NP_071301.2 | 10, 11 | 1477 | ||
| GenBank mRNA | AB049355.1 | - | 10, 11 | 1317 | ||
| AK029847.1 | - | 6, 7 | 789 | |||
| AK136597.1 | - | 10, 11 | 1387 | |||
| AK170193.1 | - | 10, 11 | 1400 | |||
| AK170347.1 | - | 1, 2 | 72 | |||
| BC021332.1 | - | 10, 11 | 1365 | |||
| BC038006.1 | - | 10, 11 | 1333 | |||
| BC058234.1 | - | 8, 9 | 1077 | 6306849 | ||
| BC099690.1 | - | 10, 11 | 1384 | |||
| BC137843.1 | - | 6, 7 | 854 | 8860862 | ||

Fam129a
Genomic Map