siRNA Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022019.5 | NP_071302.2 | 2 | 164 | ||
| GenBank mRNA | AB037908.1 | - | 2 | 170 | ||
| AK011995.1 | - | 2 | 148 | |||
| AK035293.1 | - | 2 | 162 | |||
| AK050528.1 | - | 2 | 162 | |||
| AK088024.1 | - | 2 | 163 | |||
| AK088186.1 | - | 2 | 165 | |||
| AK088357.1 | - | 2 | 160 | |||
| AK142568.1 | - | 2 | 166 | |||
| BC025066.1 | - | 2 | 182 | 3495358 | ||

Dusp10
Genomic Map