siRNA Details
| Gene Symbol: Ube4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022022.3 | NP_071305.2 | 23 | 3583 | ||
| GenBank mRNA | AF260926.1 | - | 23 | 3698 | ||
| AK029914.1 | - | 23 | 3188 | |||
| AK083409.1 | - | 23 | 3187 | |||
| AK122345.1 | - | 23 | 3094 | |||
| AK163319.1 | - | 11 | 1380 | |||
| AK180753.1 | - | 1 | 31 | |||
| BC067402.1 | - | 23 | 3330 | 30099547 | ||
| BC075620.1 | - | 23 | 3544 | 6405514 | ||

Ube4b
Genomic Map