siRNA Details
| Gene Symbol: Slc5a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022025.4 | NP_071308.2 | 2 | 547 | ||
| GenBank mRNA | AF276872.2 | - | 1 | 38 | ||
| AJ401467.1 | - | 1 | 38 | |||
| AK034415.1 | - | 2 | 431 | |||
| AK053063.1 | - | 2 | 549 | |||
| BC065089.1 | - | 1 | 64 | 6851099 | ||

Slc5a7
Genomic Map