siRNA Details
| Gene Symbol: Sv2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022030.3 | NP_071313.1 | 4 | 1339 | ||
| GenBank mRNA | AK028318.1 | - | 4 | 1284 | ||
| AK080502.1 | - | 4 | 1292 | |||
| AK080525.1 | - | 4 | 1381 | |||
| AK122360.1 | - | 4 | 1375 | |||
| AK144166.1 | - | 4 | 1362 | |||
| BC046587.1 | - | 4 | 1339 | 6493509 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK158936.1 | - | 2803 | Chr. - - [-] on Build | ||

Sv2a
Genomic Map