siRNA Details
| Gene Symbol: Parvg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162500.1 | NP_001155972.1 | 11 | 942 | ||
| NM_022321.4 | NP_071716.3 | 11 | 897 | |||
| GenBank mRNA | AF312712.1 | - | 11 | 855 | ||
| AK028329.1 | - | 11 | 898 | |||
| AK037365.1 | - | 11 | 870 | |||
| AK040914.1 | - | 11 | 828 | |||
| AK079881.1 | - | 11 | 823 | |||
| AK088419.1 | - | 11 | 847 | |||

Parvg
Genomic Map