siRNA Details
| Gene Symbol: Mllt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022328.2 | NP_071723.1 | 2 | 204 | ||
| GenBank mRNA | AF298887.1 | - | 1 | 90 | ||
| AF312858.1 | - | 2 | 101 | |||
| AK144605.1 | - | 2 | 232 | |||
| AK156820.1 | - | 2 | 231 | |||
| BC053005.1 | - | 2 | 204 | 5716396 | ||

Mllt1
Genomic Map