siRNA Details
| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207530.3 | NP_997413.2 | 13 | 1287 | ||
| GenBank mRNA | AY536214.1 | - | 13 | 1166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osbpl1a| Gene Symbol: Osbpl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207530.3 | NP_997413.2 | 13 | 1287 | ||
| GenBank mRNA | AY536214.1 | - | 13 | 1166 | ||