siRNA Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122765.1 | NP_001116237.1 | 5 | 412 | ||
| NM_001122766.1 | NP_001116238.1 | 3 | 249 | |||
| NM_022432.4 | NP_071877.3 | 6 | 459 | |||
| GenBank mRNA | AB221570.1 | - | 6 | 364 | ||
| AF299337.1 | - | 6 | 444 | |||
| AK014042.1 | - | 5 | 383 | |||
| AK144431.1 | - | 5 | 355 | |||
| AK146412.1 | - | 5 | 392 | |||
| BB868649.1 | - | 3 | 214 | |||
| BC021439.1 | - | 6 | 463 | 5039278 | ||
| KF032392.1 | - | 6 | 401 | |||

Sirt2
Genomic Map