siRNA Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122765.1 | NP_001116237.1 | 11 | 815 | ||
| NM_001122766.1 | NP_001116238.1 | 9 | 652 | |||
| NM_022432.4 | NP_071877.3 | 12 | 862 | |||
| GenBank mRNA | AB221570.1 | - | 12 | 767 | ||
| AF299337.1 | - | 12 | 847 | |||
| AK014042.1 | - | 11 | 786 | |||
| AK144431.1 | - | 11 | 759 | |||
| AK146412.1 | - | 11 | 792 | |||
| AK180132.1 | - | 4 | 297 | |||
| AK183534.1 | - | 4 | 236 | |||
| BC021439.1 | - | 12 | 866 | 5039278 | ||

Sirt2
Genomic Map