siRNA Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127351.1 | NP_001120823.1 | 3 | 321 | ||
| NM_001177804.1 | NP_001171275.1 | 3 | 313 | |||
| NM_022433.2 | NP_071878.2 | 3 | 366 | |||
| GenBank mRNA | AF299338.1 | - | 3 | 366 | ||
| AF299339.1 | - | 3 | 321 | |||
| AK075861.1 | - | 3 | 295 | |||
| BC025878.1 | - | 3 | 265 | 5253854 | ||
| CB953804.1 | - | 3 | 313 | |||
| CT010402.1 | - | 2 | 72 | |||
| EU886466.1 | - | 3 | 311 | |||
| FJ621493.1 | - | 3 | 309 | |||

Sirt3
Genomic Map