siRNA Details
| Gene Symbol: Polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285800.1 | NP_001272729.1 | 10 | 953 | ||
| NM_022811.3 | NP_073722.1 | 8 | 809 | |||
| GenBank mRNA | AK050780.1 | - | 757 | |||
| AK083948.1 | - | 8 | 779 | |||
| AK132556.1 | - | 8 | 780 | |||
| BC034783.1 | - | 10 | 900 | 4022372 | ||
| D14336.1 | - | 8 | 772 | |||

Polr1e
Genomic Map