siRNA Details
| Gene Symbol: Fcamr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170632.1 | NP_001164103.1 | 4 | 542 | ||
| NM_144960.2 | NP_659209.2 | 4 | 374 | |||
| GenBank mRNA | AB048834.1 | - | 4 | 386 | ||
| BC110549.1 | - | 4 | 545 | 40047147 | ||
| BC110550.1 | - | 4 | 377 | 40047153 | ||
| FJ011703.1 | - | 3 | 455 | |||

Fcamr
Genomic Map