siRNA Details
| Gene Symbol: Gpr85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145066.4 | NP_659503.1 | 4 | 1606 | ||
| GenBank mRNA | AF254416.2 | - | 1 | 642 | ||
| AK043090.1 | - | 2 | 831 | |||
| AK047740.1 | - | 3 | 809 | |||
| BC026975.1 | - | 1 | 823 | 5363257 | ||
| BC036271.1 | - | 1 | 823 | |||
| BC065154.1 | - | 4 | 1606 | 6418997 | ||
More Information
Set Membership: |
|

Gpr85
Genomic Map