siRNA Details
Gene Symbol: Slc5a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133184.2 | NP_573447.2 | 1, 2 | 218 | ||
GenBank mRNA | AF251267.1 | - | 1, 2 | 218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc5a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133184.2 | NP_573447.2 | 1, 2 | 218 | ||
GenBank mRNA | AF251267.1 | - | 1, 2 | 218 |