siRNA Details
| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022565.2 | NP_072087.2 | 10 | 2631 | ||
| GenBank mRNA | AB036838.1 | - | 9 | 2081 | ||
| AK132938.1 | - | 5 | 786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndst4| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022565.2 | NP_072087.2 | 10 | 2631 | ||
| GenBank mRNA | AB036838.1 | - | 9 | 2081 | ||
| AK132938.1 | - | 5 | 786 | |||