siRNA Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 5 | 711 | ||
| GenBank mRNA | AF016402.1 | - | 1 | 171 | ||
| BC044665.1 | - | 5 | 678 | 2647641 | ||
| CN534597.1 | - | 3 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ireb2| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 5 | 711 | ||
| GenBank mRNA | AF016402.1 | - | 1 | 171 | ||
| BC044665.1 | - | 5 | 678 | 2647641 | ||
| CN534597.1 | - | 3 | 365 | |||