siRNA Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 9 | 1180 | ||
| GenBank mRNA | BC044665.1 | - | 9 | 1147 | 2647641 | |
| CJ093933.1 | - | 5 | 568 | |||
| CK636500.1 | - | 5 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ireb2| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 9 | 1180 | ||
| GenBank mRNA | BC044665.1 | - | 9 | 1147 | 2647641 | |
| CJ093933.1 | - | 5 | 568 | |||
| CK636500.1 | - | 5 | 472 | |||