siRNA Details
| Gene Symbol: Keg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029550.4 | NP_083826.1 | 6 | 911 | ||
| GenBank mRNA | AB028071.1 | - | 6 | 887 | ||
| AK002360.1 | - | 6 | 909 | |||
| AK165515.1 | - | 6 | 909 | |||
| BC010803.1 | - | 6 | 901 | 4238654 | ||

Keg1
Genomic Map