siRNA Details
| Gene Symbol: Lpin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164885.1 | NP_001158357.1 | 7 | 1358 | ||
| NM_022882.4 | NP_075020.2 | 7 | 1234 | |||
| GenBank mRNA | AF286723.1 | - | 7 | 1234 | ||
| AK033662.1 | - | 7 | 1271 | |||
| AK136472.1 | - | 1829 | ||||
| BC039698.1 | - | 7 | 1347 | 5101211 | ||

Lpin2
Genomic Map