siRNA Details
| Gene Symbol: Scel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022886.2 | NP_075024.2 | 3 | 219 | ||
| GenBank mRNA | AF245700.2 | - | 3 | 219 | ||
| AK020346.1 | - | 3 | 141 | |||
| BC025142.1 | - | 3 | 197 | |||
| BC111874.1 | - | 3 | 139 | 40059586 | ||
| BC112380.1 | - | 3 | 140 | 40059585 | ||
| BY775767.1 | - | 3 | 220 | |||

Scel
Genomic Map