siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 11 | 1354 | ||
GenBank mRNA | AF226323.1 | - | 11 | 1373 | ||
AK135680.1 | - | 11 | 1660 | |||
AK142382.1 | - | 11 | 1641 | |||
AK145421.1 | - | 10 | 1323 | |||
AK150682.1 | - | 11 | 1368 | |||
AK162726.1 | - | 11 | 1333 | |||
AK166821.1 | - | 11 | 1347 | |||
AK167421.1 | - | 11 | 1371 | |||
AK167594.1 | - | 11 | 1367 | |||
AK168880.1 | - | 11 | 1347 | |||
AK170473.1 | - | 11 | 1388 | |||
AK208279.1 | - | 2 | 211 | |||
AK210202.1 | - | 2 | 210 | |||
BC005469.1 | - | 11 | 1379 | 3492842 | ||
BC006637.1 | - | 11 | 1359 | 3969440 | ||
U47024.1 | - | 10 | 1328 |
