siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 10 | 1076 | ||
GenBank mRNA | AF226323.1 | - | 10 | 1095 | ||
AK135680.1 | - | 10 | 1382 | |||
AK142382.1 | - | 10 | 1363 | |||
AK145421.1 | - | 9 | 1045 | |||
AK145853.1 | - | 10 | 1125 | |||
AK150682.1 | - | 10 | 1090 | |||
AK162726.1 | - | 10 | 1055 | |||
AK166821.1 | - | 10 | 1069 | |||
AK167421.1 | - | 10 | 1093 | |||
AK167594.1 | - | 10 | 1089 | |||
AK168880.1 | - | 10 | 1069 | |||
AK170473.1 | - | 10 | 1110 | |||
AK190441.1 | - | 2 | 89 | |||
BC005469.1 | - | 10 | 1101 | 3492842 | ||
BC006637.1 | - | 10 | 1081 | 3969440 | ||
U47024.1 | - | 9 | 1044 |
