siRNA Details
| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039126.2 | NP_001034215.1 | 4 | 761 | ||
| NM_023046.4 | NP_075533.3 | 4 | 562 | |||
| GenBank mRNA | AF155352.1 | - | 4 | 513 | ||
| AK006515.1 | - | 4 | 563 | |||
| AK122456.1 | - | 4 | 617 | |||
| AK153774.1 | - | 4 | 783 | |||
| BC034869.1 | - | 5 | 556 | |||
| BC049699.1 | - | 4 | 562 | |||
| BU938650.1 | - | 4 | 565 | |||
| KC789642.1 | - | 3 | 291 | |||

Asb1
Genomic Map