siRNA Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023048.5 | NP_075535.1 | 3 | 310 | ||
| GenBank mRNA | AF155355.1 | - | 2 | 280 | ||
| AK018373.1 | - | 2 | 463 | |||
| AK076004.1 | - | 2 | 365 | |||
| AK132068.1 | - | 2 | 472 | |||
| AK133894.1 | - | 2 | 181 | |||
| BC046819.1 | - | 3 | 310 | 5703017 | ||
| BC054745.1 | - | 2 | 418 | 6836490 | ||
| BC056440.1 | - | 3 | 310 | 5703017 | ||

Asb4
Genomic Map