siRNA Details
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164791.1 | NP_001158263.1 | 9 | 893 | ||
NM_023057.5 | NP_075544.1 | 9 | 957 | |||
NM_178084.4 | NP_835185.2 | 9 | 893 | |||
GenBank mRNA | AB049731.1 | - | 8 | 740 | ||
AB049732.1 | - | 8 | 740 | |||
AK045444.1 | - | 9 | 818 | |||
AK087363.1 | - | 9 | 833 | |||
AK132186.1 | - | 9 | 958 | |||
AK162819.1 | - | 9 | 758 | |||
BC023718.1 | - | 9 | 838 | 5346593 |
