siRNA Details
| Gene Symbol: Cubn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081084.2 | NP_001074553.1 | 51 | 8108 | ||
| GenBank mRNA | CF169457.1 | - | 3 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cubn| Gene Symbol: Cubn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081084.2 | NP_001074553.1 | 51 | 8108 | ||
| GenBank mRNA | CF169457.1 | - | 3 | 428 | ||