siRNA Details
| Gene Symbol: Lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113545.1 | NP_001107017.1 | 11 | 1788 | ||
| NM_023063.4 | NP_075550.2 | 8 | 1660 | |||
| GenBank mRNA | AF307844.1 | - | 8 | 1583 | ||
| AF307845.1 | - | 11 | 1742 | |||
| AK028186.1 | - | 8 | 1601 | |||
| AK031698.1 | - | 8 | 1604 | |||
| AK049350.1 | - | 11 | 1738 | |||
| AK085065.1 | - | 8 | 1654 | |||
| AK132314.1 | - | 8 | 1606 | |||
| AK167220.1 | - | 8 | 1220 | |||
| BC031490.1 | - | 11 | 1714 | 3498138 | ||

Lima1
Genomic Map