siRNA Details
Gene Symbol: Gtpbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159329.1 | NP_001152801.1 | 1 | 149 | ||
NM_025332.3 | NP_079608.1 | 1 | 149 | |||
GenBank mRNA | AK002771.1 | - | 1 | 134 | ||
AK010979.1 | - | 1 | 146 | |||
AK044298.1 | - | 346 | ||||
AK078242.1 | - | 1 | 150 | |||
AK147840.1 | - | 1 | 133 | |||
BC107216.1 | - | 1 | 146 | 40051675 | ||
BC107217.1 | - | 1 | 146 | 40051680 | ||
CJ169587.1 | - | 1 | 150 |
