siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159329.1 | NP_001152801.1 | 3 | 354 | ||
| NM_025332.3 | NP_079608.1 | 3 | 555 | |||
| GenBank mRNA | AK002771.1 | - | 3 | 540 | ||
| AK010979.1 | - | 3 | 552 | |||
| AK147840.1 | - | 3 | 539 | |||
| BC107216.1 | - | 3 | 552 | 40051675 | ||
| BC107217.1 | - | 3 | 351 | 40051680 | ||

Gtpbp8
Genomic Map