siRNA Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025333.4 | NP_079609.2 | 2, 3 | 374 | ||
| GenBank mRNA | AK013454.1 | - | 2, 3 | 279 | ||
| AK075594.1 | - | 2, 3 | 279 | |||
| AK160957.1 | - | 2, 3 | 281 | |||
| AK167228.1 | - | 2, 3 | 310 | |||
| BC029630.1 | - | 1, 2 | 93 | |||
| BC058955.1 | - | 2, 3 | 323 | |||
| BC060090.1 | - | 2, 3 | 323 | 5698361 | ||
| BC085277.1 | - | 2, 3 | 276 | 30441476 | ||

Sdhaf2
Genomic Map