siRNA Details
| Gene Symbol: Ghitm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199122.1 | NP_001186051.1 | 3 | 419 | ||
| NM_078478.5 | NP_510963.1 | 3 | 372 | |||
| GenBank mRNA | AF412297.1 | - | 3 | 316 | ||
| AK005636.1 | - | 3 | 316 | |||
| AK093887.1 | - | 3 | 348 | |||
| AK134592.1 | - | 3 | 302 | |||
| AK146433.1 | - | 3 | 279 | |||
| AK151488.1 | - | 3 | 349 | |||
| AK158904.1 | - | 3 | 362 | |||
| AK159695.1 | - | 3 | 346 | |||
| AK159724.1 | - | 3 | 346 | |||
| AK164122.1 | - | 3 | 317 | |||
| AK167970.1 | - | 3 | 343 | |||
| AK168022.1 | - | 3 | 349 | |||
| AK168678.1 | - | 3 | 420 | |||
| BC008622.1 | - | 3 | 350 | 3588758 | ||
| BC010224.1 | - | 3 | 352 | 3500685 | ||

Ghitm
Genomic Map