siRNA Details
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 9 | 1239 | ||
NM_078478.5 | NP_510963.1 | 9 | 1192 | |||
GenBank mRNA | AF412297.1 | - | 9 | 1136 | ||
AK005636.1 | - | 9 | 1137 | |||
AK093887.1 | - | 9 | 1168 | |||
AK134592.1 | - | 9 | 1122 | |||
AK146433.1 | - | 9 | 1099 | |||
AK146853.1 | - | 9 | 1155 | |||
AK151488.1 | - | 9 | 1169 | |||
AK158904.1 | - | 9 | 1182 | |||
AK159695.1 | - | 9 | 1166 | |||
AK159724.1 | - | 9 | 1166 | |||
AK164122.1 | - | 9 | 1137 | |||
AK167970.1 | - | 9 | 1163 | |||
AK168022.1 | - | 9 | 1169 | |||
AK168678.1 | - | 9 | 1240 | |||
AK168800.1 | - | 9 | 1148 | |||
AK210138.1 | - | 2 | 122 | |||
BC008622.1 | - | 9 | 1170 | 3588758 | ||
BC010224.1 | - | 9 | 1172 | 3500685 |
