siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 6 | 1135 | ||
| GenBank mRNA | AK138874.1 | - | 1 | 783 | ||
| AY364165.1 | - | 6 | 920 | |||
| AY846873.1 | - | 6 | 1136 | |||
| AY846874.1 | - | 6 | 1136 | |||
| BC019186.1 | - | 6 | 1104 | |||
| BC043306.1 | - | 3 | 567 | |||
| BC054355.1 | - | 7 | 1048 | |||
| BC054769.1 | - | 6 | 1028 | 6415086 | ||

Tmem57
Genomic Map