siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 2 | 425 | ||
| GenBank mRNA | AK038972.1 | - | 2 | 319 | ||
| AY364165.1 | - | 2 | 210 | |||
| AY846873.1 | - | 2 | 426 | |||
| AY846874.1 | - | 2 | 426 | |||
| BC010714.1 | - | 2 | 274 | |||
| BC019186.1 | - | 2 | 394 | |||
| BC037192.1 | - | 2 | 394 | |||
| BC042733.2 | - | 2 | 365 | |||
| BC044802.1 | - | 2 | 365 | |||
| BC054355.1 | - | 2 | 229 | |||
| BC054769.1 | - | 2 | 318 | 6415086 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001025699, XM_008764241, AY845021, BC097999.

Tmem57
Genomic Map