siRNA Details
| Gene Symbol: Fbxo36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025386.3 | NP_079662.1 | 1, 2 | 127 | ||
| GenBank mRNA | AK003856.1 | - | 1, 2 | 127 | ||
| AK008324.1 | - | 1, 2 | 124 | |||
| BC049581.1 | - | 1, 2 | 104 | 6772363 | ||
| BY708252.1 | - | 1, 2 | 124 | |||

Fbxo36
Genomic Map