siRNA Details
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294269.1 | NP_001281198.1 | 5 | 1074 | ||
| NM_025396.3 | NP_079672.1 | 5 | 790 | |||
| GenBank mRNA | AK003134.1 | - | 5 | 787 | ||
| AK003985.1 | - | 5 | 791 | |||
| AK172066.1 | - | 5 | 791 | |||
| AK188109.1 | - | 2 | 210 | |||

Pgls
Genomic Map