siRNA Details
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025396.3 | NP_079672.1 | 1, 2 | 324 | ||
| GenBank mRNA | AK003134.1 | - | 1, 2 | 321 | ||
| AK003985.1 | - | 1, 2 | 325 | |||
| AK036059.1 | - | 325 | ||||
| AK172066.1 | - | 1, 2 | 325 | |||
| BC006594.1 | - | 1, 2 | 298 | 3964951 | ||

Pgls
Genomic Map