siRNA Details
| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 21 | 4845 | ||
| GenBank mRNA | AK122509.1 | - | 20 | 4598 | ||
| BC028830.1 | - | 3 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110037F02Rik| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 21 | 4845 | ||
| GenBank mRNA | AK122509.1 | - | 20 | 4598 | ||
| BC028830.1 | - | 3 | 352 | |||