siRNA Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290660.1 | NP_001277589.1 | 3 | 585 | ||
| NM_025422.4 | NP_079698.2 | 3 | 585 | |||
| GenBank mRNA | AI116405.1 | - | 3 | 585 | ||
| AK002812.1 | - | 3 | 226 | |||
| AK004267.1 | - | 3 | 521 | |||
| BC005501.2 | - | 2 | 147 | 3489789 | ||

Cd302
Genomic Map