siRNA Details
Gene Symbol: Ndufb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023172.3 | NP_075661.1 | 2 | 267 | ||
GenBank mRNA | AK002254.1 | - | 2 | 268 | ||
AK003540.1 | - | 2 | 268 | |||
AK004522.1 | - | 2 | 268 | |||
BC031539.1 | - | 2 | 249 | 3979845 |
